Benzotrichloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15652-0E
A15652-36
CAAAA15652-0EEA
1085.46
CAD
CAAAA15652-0E
CAAAA15652-36
Benzotrichloride 98%
Benzotrichloride
Purity: 98%
CAS No.: 98-07-7
Molecular Formula: C7H5Cl3
Formula Weight: 195.48
Flash Point: 97°(207°F)
Refractive Index: 1.5570
Melting Point: -8 to -.5°
Boiling Point: 90-91°/15mm
Beilstein Reference: 508152
Merck Reference: 14,1109
UN No.: 2226
MDL No.: MFCD00000786
Formula:
C₇H₅Cl₃ Boiling Pt: 90…91 °C/15 mm Melting Pt: –8 to –.5 °C Density: 1.38 Flash Pt: 97 °C (207 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000786 CAS Number: 98-07-7 EINECS: 202-634-5 UN: 2226 ADR: 8,II Merck Index: 14,1109 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...