2,3-Diaminotoluene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Methylbenzene-1,2-diamine, Toluene-2,3-diamine, 1,2-Diamino-3-methylbenzene
A15647-06
A15647-14
CAAAA15647-06EA
135.88
CAD
CAAAA15647-06
CAAAA15647-14
2,3-Diaminotoluene 97%
2,3-Diaminotoluene
Purity: 97%
CAS No.: 2687-25-4
Molecular Formula: C7H10N2
Formula Weight: 122.17
Melting Point: 56-60°
Boiling Point: 225°
MDL No.: MFCD00011589
Formula:
C₇H₁₀N₂ MW: 122.17 g/mol Boiling Pt: 225 °C Melting Pt: 56…60 °C Density: 1.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011589 CAS Number: 2687-25-4 EINECS: 220-248-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...