(Methyl)triphenylphosphonium iodide 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyltriphenylphosphonium iodide

A15644-14 A15644-22
CAAAA15644-14EA 100.65 CAD
CAAAA15644-14 CAAAA15644-22 CAAAA15644-36
(Methyl)triphenylphosphonium iodide 98%
(Methyl)triphenylphosphonium iodide

Purity: 98% CAS No.: 2065-66-9 Molecular Formula: CH3P(C6H5)3I Formula Weight: 404.23 Odor: Odorless Melting Point: 185-188° UN No.: 3464 MDL No.: MFCD00066175

Formula: C₁₉H₁₈IP
MW: 404.23 g/mol
Melting Pt: 185…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00066175
CAS Number: 2065-66-9
EINECS: 218-178-5
UN: 3464
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR