(3-Fluorophenyl)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(3-Fluorophenyl)acetic acid

A15635-06 A15635-14 A15635-22
CAAAA15635-06EA 93.54 CAD
CAAAA15635-06 CAAAA15635-14 CAAAA15635-22
(3-Fluorophenyl)acetic acid 98%
(3-Fluorophenyl)acetic acid

Purity: 98% CAS No.: 331-25-9 Molecular Formula: C8H7FO2 Formula Weight: 154.14 Flash Point: 110°(230°F) Odor: Odorless Boiling Point: 151°/0.8mm Beilstein Reference: 775898 MDL No.: MFCD00004331

Formula: (C₆H₄F)CH₂COOH
MW: 154.14 g/mol
Boiling Pt: 151 °C (0.8 mmHg)
Melting Pt: 44…48 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004331
CAS Number: 331-25-9
EINECS: 206-360-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR