4-Bromobutyl acetate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Acetic acid 4-bromobutyl ester
A15632-09
A15632-18
CAAAA15632-09EA
95.27
CAD
CAAAA15632-09
CAAAA15632-18
CAAAA15632-30
4-Bromobutyl acetate 98+%
4-Bromobutyl acetate
Purity: 98+%
CAS No.: 4753-59-7
Molecular Formula: CH3CO2(CH2)4Br
Formula Weight: 195.07
Flash Point: 109°(228°F)
Refractive Index: 1.4600
Boiling Point: 96-97°/15mm
Beilstein Reference: 1749695
MDL No.: MFCD00000263
Formula:
C₆H₁₁BrO₂ MW: 195.06 Boiling Pt: 96…97 °C (15 mmHg) Density: 1.348 Flash Pt: 109 °C (228 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000263 CAS Number: 4753-59-7 EINECS: 225-277-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...