4-Iodotoluene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15631-14
A15631-22
A15631-36
CAAAA15631-14EA
66.99
CAD
CAAAA15631-14
CAAAA15631-22
CAAAA15631-36
4-Iodotoluene 98%
4-Iodotoluene
Purity: 98%
CAS No.: 624-31-7
Molecular Formula: C7H7I
Formula Weight: 218.04
Flash Point: 90°(194°F)
Melting Point: 33-35°
Boiling Point: 210-212°
Beilstein Reference: 1903637
MDL No.: MFCD00001059
Formula:
C₇H₇I Boiling Pt: 210…212 °C Melting Pt: 33…35 °C Density: 1.678 Flash Pt: 90 °C(194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001059 CAS Number: 624-31-7 EINECS: 210-841-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...