Chlormequat chloride, (max. 5% H₂O) 98% (dry weight)
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15630-06
A15630-22
CAAAA15630-06EA
44.08
CAD
CAAAA15630-06
CAAAA15630-22
Chlormequat chloride, (max. 5% H₂O) 98% (dry weight)
Chlormequat chloride
Purity: 98%
CAS No.: 999-81-5
Molecular Formula: C5H12ClN.HCl
Formula Weight: 158.07
Odor: Amine-like
Melting Point: ca 241° dec.
Beilstein Reference: 3563994
Merck Reference: 14,2104
UN No.: 2811
MDL No.: MFCD00011869
Formula:
ClCH₂CH₂N(CH₃)₃Cl Melting Pt: ∼ 241 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011869 CAS Number: 999-81-5 EINECS: 213-666-4 UN: 2811 ADR: 6.1,III Merck Index: 14,2104 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...