3-Bromoanisole 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15625-14
A15625-22
A15625-36
CAAAA15625-14EA
69.99
CAD
CAAAA15625-14
CAAAA15625-22
CAAAA15625-36
3-Bromoanisole 98+%
3-Bromoanisole
Purity: 98+%
CAS No.: 2398-37-0
Molecular Formula: C7H7BrO
Formula Weight: 187.04
Flash Point: 93°(199°F)
Refractive Index: 1.5640
Melting Point: ca 2°
Boiling Point: 210-211°
Beilstein Reference: 1524687
MDL No.: MFCD00000081
Formula:
C₇H₇BrO Boiling Pt: 210…211 °C Melting Pt: ∼ 2 °C Density: 1.489 Flash Pt: 93 °C (199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000081 CAS Number: 2398-37-0 EINECS: 219-264-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...