2-Fluorobenzylamine 97%

Supplier: Thermo Scientific Chemicals
Danger

A15623-06 A15623-09 A15623-18
CAAAAA15623-06EA 105.7 CAD
CAAAAA15623-06 CAAAA15623-09 CAAAA15623-18
2-Fluorobenzylamine 97%
2-Fluorobenzylamine

Purity: 97% CAS No.: 89-99-6 Molecular Formula: C7H8FN Formula Weight: 125.15 Flash Point: 67°(152°F) Refractive Index: 1.5170 Odor: Amine-like Boiling Point: 80-82°/18mm Beilstein Reference: 508109 UN No.: 2735 MDL No.: MFCD00008107

Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 80…82 °C/18 mm
Density: 1.099
Flash Pt: 67 °C (152 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008107
CAS Number: 89-99-6
EINECS: 201-957-9
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR