6-Chloronicotinic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15620-06
A15620-14
A15620-22
CAAAA15620-06EA
65.41
CAD
CAAAA15620-06
CAAAA15620-14
CAAAA15620-22
6-Chloronicotinic acid 99%
6-Chloronicotinic acid
Purity: 99%
CAS No.: 5326-23-8
Molecular Formula: C6H4ClNO2
Formula Weight: 157.56
Melting Point: ca 192° dec.
Beilstein Reference: 115993
Formula:
C₆H₄ClNO₂ Boiling Pt: 284 °C (1013 hPa) Melting Pt: ∼ 192 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00006241 CAS Number: 5326-23-8 EINECS: 226-201-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...