4-tert-Butylcyclohexanone 99%

Supplier: Thermo Scientific Chemicals
Warning

A15612-22 A15612-36
CAAAA15612-22EA 111.39 CAD
CAAAA15612-22 CAAAA15612-36
4-tert-Butylcyclohexanone 99%
4-tert-Butylcyclohexanone

Purity: 99% CAS No.: 98-53-3 Molecular Formula: C10H18O Formula Weight: 154.25 Flash Point: 96°(204°F) Melting Point: 47-51° Boiling Point: 113-116°/20mm Beilstein Reference: 507309 MDL No.: MFCD00001642

Formula: C₁₀H₁₈O
Boiling Pt: 113…116 °C/20 mm
Melting Pt: 47…51 °C
Density: 0.893
Flash Pt: 96 °C(204 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001642
CAS Number: 98-53-3
EINECS: 202-678-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR