Laurinaldehyde 95% stabilised
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Lauric aldehyde, Dodecylaldehyde, Dodecanal
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) ≤ 250 ppm
A15611-22
A15611-36
CAAAA15611-22EA
123.4
CAD
CAAAA15611-22
CAAAA15611-36
Laurinaldehyde 95% stabilised
Laurinaldehyde
Purity: 95%
CAS No.: 112-54-9
Molecular Formula: CH3(CH2)10CHO
Formula Weight: 184.32
Flash Point: 101°(214°F)
Refractive Index: 1.435
Melting Point: 12°
Boiling Point: 184-186°/100mm
Beilstein Reference: 1703917
MDL No.: MFCD00007017
Formula:
C₁₂H₂₄O Boiling Pt: 184…186 °C/100 mm Melting Pt: 12 °C Density: 0.835 Flash Pt: 101 °C (214 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007017 CAS Number: 112-54-9 EINECS: 203-983-6 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...