Cyclohexanone 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15607-0F
A15607-AP
CAAAAA15607-AEEA
45.05
CAD
CAAAAA15607-AE
CAAAA15607-0F
CAAAA15607-AP
Cyclohexanone 99+%
Cyclohexanone
Purity: 99+%
CAS No.: 108-94-1
Molecular Formula: C6H10O
Formula Weight: 98.15
Flash Point: 47°(116°F)
Refractive Index: 1.4507
Melting Point: -26°
Boiling Point: 154-156°
Beilstein Reference: 385735
Merck Reference: 14,2726
UN No.: 1915
MDL No.: MFCD00001625
Formula:
C₆H₁₀(=O) Boiling Pt: 154…156 °C Melting Pt: –26 °C Density: 0.948 Flash Pt: 47 °C (116 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001625 CAS Number: 108-94-1 EINECS: 203-631-1 UN: 1915 ADR: 3,III REACH: 01-2119453616-35 Merck Index: 14,2726 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...