3'-Methoxyacetophenone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15604-36
A15604-14
A15604-22
CAAAAA15604-36EA
571.96
CAD
CAAAAA15604-36
CAAAA15604-14
CAAAA15604-22
3'-Methoxyacetophenone 97%
3'-Methoxyacetophenone
Purity: 97%
CAS No.: 586-37-8
Molecular Formula: C9H10O2
Formula Weight: 150.18
Flash Point:
Formula:
C₉H₁₀O₂ Boiling Pt: 239…241 °C Melting Pt: –7 °C Density: 1.094 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008736 CAS Number: 586-37-8 EINECS: 209-573-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...