4-Chlorophenol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
p-Chlorophenol
A15602-22
A15602-0B
A15602-30
CAAAAA15602-22EA
65.73
CAD
CAAAAA15602-22
CAAAA15602-0B
CAAAA15602-30
4-Chlorophenol 99%
4-Chlorophenol
Purity: 99%
CAS No.: 106-48-9
Molecular Formula: C6H5ClO
Formula Weight: 128.56
Flash Point: 115°(239°F)
Odor: Phenol-like
Boiling Point: 217-220°
Beilstein Reference: 507004
Merck Reference: 14,2154
UN No.: 2020
MDL No.: MFCD00002318
Formula:
C₆H₅ClO MW: 128.56 g/mol Boiling Pt: 217…220 °C Melting Pt: 39…43 °C Density: 1.306 Flash Pt: 115 °C (239 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002318 CAS Number: 106-48-9 EINECS: 203-402-6 UN: 2020 ADR: 6.1,III Merck Index: 14,2154 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...