3,4-Difluorobenzonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15600-06
A15600-14
A15600-22
CAAAA15600-06EA
90.69
CAD
CAAAA15600-06
CAAAA15600-14
CAAAA15600-22
3,4-Difluorobenzonitrile 98%
3,4-Difluorobenzonitrile
Purity: 98%
CAS No.: 64248-62-0
Molecular Formula: C7H3F2N
Formula Weight: 139.11
Flash Point: 69°(156°F)
Melting Point: 48-52°
Boiling Point: 180°
Beilstein Reference: 2082224
UN No.: 3439
MDL No.: MFCD00011666
Formula:
C₇H₃F₂N Boiling Pt: 180 °C Melting Pt: 48…52 °C Flash Pt: 69 °C (156 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011666 CAS Number: 64248-62-0 EINECS: 264-751-8 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...