4-Fluoroaniline 99%

Supplier: Thermo Scientific Chemicals
Danger

A15597-14 A15597-22 A15597-36
CAAAA15597-14EA 42.03 CAD
CAAAA15597-14 CAAAA15597-22 CAAAA15597-36
4-Fluoroaniline 99%
4-Fluoroaniline

Purity: 99% CAS No.: 371-40-4 Molecular Formula: C6H6FN Formula Weight: 111.12 Flash Point: 73°(163°F) Refractive Index: 1.5400 Odor: Recognizable Melting Point: -2° Boiling Point: 186-187° Beilstein Reference: 742030 Merck Reference: 14,4169 UN No.: 2941 MDL No.: MFCD00007829

Formula: C₆H₆FN
MW: 111.12 g/mol
Boiling Pt: 186…187 °C
Melting Pt: –2 °C
Density: 1.158
Flash Pt: 73 °C (163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007829
CAS Number: 371-40-4
EINECS: 206-735-5
UN: 2941
ADR: 6.1,III
Merck Index: 14,4169

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR