4-Picoline 98%

Supplier: Thermo Scientific Chemicals
Danger

A15595-0F A15595-AE A15595-AP
CAAAA15595-0FEA 200.66 CAD
CAAAA15595-0F CAAAA15595-AE CAAAA15595-AP
4-Picoline 98%
4-Picoline

Purity: 98% CAS No.: 108-89-4 Molecular Formula: C6H7N Formula Weight: 93.13 Flash Point: 39°(102°F) Refractive Index: 1.5050 Odor: Unpleasant pH: 10-11 Melting Point: 3° Boiling Point: 144-145° Beilstein Reference: 104586 Merck Reference: 14,7402 UN No.: 2313 MDL No.: MFCD00006440

Formula: C₆H₇N
Boiling Pt: 144…145 °C
Melting Pt: 3 °C
Density: 0.957
Flash Pt: 39 °C (102 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006440
CAS Number: 108-89-4
EINECS: 203-626-4
UN: 1992
ADR: 3,III
Merck Index: 14,7402

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR