2-Chlorobenzylamine 96%

Supplier: Thermo Scientific Chemicals
Danger

A15594-36 A15594-14 A15594-22
CAAAAA15594-36EA 861.1 CAD
CAAAAA15594-36 CAAAA15594-14 CAAAA15594-22
2-Chlorobenzylamine 96%
2-Chlorobenzylamine
Formula: C₇H₈ClN
Boiling Pt: 227 °C
Density: 1.170
Flash Pt: 88 °C (190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008108
CAS Number: 89-97-4
EINECS: 201-955-8
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR