3'-Chloroacetophenone 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15592-06
A15592-14
A15592-22
CAAAA15592-06EA
57.99
CAD
CAAAA15592-06
CAAAA15592-14
CAAAA15592-22
3'-Chloroacetophenone 98+%
3'-Chloroacetophenone
Purity: 98+%
CAS No.: 99-02-5
Molecular Formula: C8H7ClO
Formula Weight: 154.60
Flash Point: 105°(221°F)
Refractive Index: 1.5506
Boiling Point: 227-229°
Beilstein Reference: 636318
UN No.: 3416
MDL No.: MFCD00000593
Formula:
C₈H₇ClO Boiling Pt: 227…229 °C Density: 1.191 Flash Pt: 105 °C(221 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000593 CAS Number: 99-02-5 EINECS: 202-721-8 UN: 3416 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...