4-Toluoyl chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: p-Toluoyl chloride, 4-Methylbenzene-1-carbonyl chloride, p-Methylbenzoyl chloride, 4-Methylbenzoyl chloride

A15581-0E A15581-22 A15581-36
CAAAA15581-0EEA 767.88 CAD
CAAAA15581-0E CAAAA15581-22 CAAAA15581-36
4-Toluoyl chloride 99%
4-Toluoyl chloride

Purity: 99% CAS No.: 874-60-2 Molecular Formula: C8H7ClO Formula Weight: 154.60 Flash Point: 82°(179°F) Physical Form: Liquid Density: 1.169 Refractive Index: 1.5530 Sensitivity: Moisture Sensitive Appearance: Colorless to pale yellow Odor: Pungent Melting Point: -2° Boiling Point: 225-227° Beilstein Reference: 471492 UN No.: 3265 MDL No.: MFCD00000696

Formula: C₈H₇ClO
MW: 154.6 g/mol
Boiling Pt: 225…227 °C
Melting Pt: –2 °C
Density: 1.169
Flash Pt: 82 °C (179 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000696
CAS Number: 874-60-2
EINECS: 212-864-8
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR