2-Phenyl-2-propanol 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: α,α-Dimethylbenzyl alcohol

A15580-14 A15580-22 A15580-36
CAAAA15580-14EA 129.88 CAD
CAAAA15580-14 CAAAA15580-22 CAAAA15580-36
2-Phenyl-2-propanol 98+%
2-Phenyl-2-propanol

Purity: 98+% CAS No.: 617-94-7 Molecular Formula: C9H12O Formula Weight: 136.19 Flash Point: 87°(188°F) Refractive Index: 1.5196 Melting Point: 29-34° Boiling Point: 215° Beilstein Reference: 1905012 MDL No.: MFCD00004456

Formula: C₆H₅C(OH)(CH₃)₂
MW: 136.19 g/mol
Boiling Pt: 215 °C
Melting Pt: 29…34 °C
Density: 0.973
Flash Pt: 87 °C(188 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004456
CAS Number: 617-94-7
EINECS: 210-539-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f8edbb976395240a99a4f52addfbfe32e8c926f&vid=5b4ae0c8b87b317b3d1326768c6ff8bdaaab37f6&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR