Pentafluorophenol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,3,4,5,6-Pentafluorophenol
A15574-06
A15574-14
A15574-22
CAAAA15574-06EA
64.15
CAD
CAAAA15574-06
CAAAA15574-14
CAAAA15574-22
Pentafluorophenol 99%
Pentafluorophenol
Purity: 99%
CAS No.: 771-61-9
Molecular Formula: C6HF5O
Formula Weight: 184.06
Flash Point: 72°(161°F)
Refractive Index: 1.4270
Melting Point: 32-36°
Boiling Point: 143°
Beilstein Reference: 1912584
MDL No.: MFCD00002156
Formula:
C₆HF₅O MW: 184.07 g/mol Boiling Pt: 143 °C Melting Pt: 32…36 °C Density: 1.757 Flash Pt: 72 °C (161 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002156 CAS Number: 771-61-9 EINECS: 212-235-8 UN: 2923 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...