Itaconic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methylenesuccinic acid, Methylenesuccinic acid

A15566-0B A15566-0I A15566-30
CAAAA15566-0BEA 81.84 CAD
CAAAA15566-0B CAAAA15566-0I CAAAA15566-30
Itaconic acid 99%
Itaconic acid

Purity: 99% CAS No.: 97-65-4 Molecular Formula: C5H6O4 Formula Weight: 130.10 Melting Point: 165-169° Boiling Point: 268° Merck Reference: 14,5242 MDL No.: MFCD00004260

Formula: C₅H₆O₄
MW: 130.1 g/mol
Boiling Pt: 268 °C
Melting Pt: 165…169 °C
Density: 1.630
Storage Temperature: Ambient
MDL Number: MFCD00004260
CAS Number: 97-65-4
EINECS: 202-599-6
Merck Index: 14,5242

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR