2,5-Dihydroxybenzaldehyde 98+%

Supplier: Thermo Scientific Chemicals
Warning

A15565-03 A15565-06 A15565-14
CAAAA15565-03EA 72.52 CAD
CAAAA15565-03 CAAAA15565-06 CAAAA15565-14
2,5-Dihydroxybenzaldehyde 98+%
2,5-Dihydroxybenzaldehyde

Purity: 98+% CAS No.: 1194-98-5 Molecular Formula: C7H6O3 Formula Weight: 138.12 Melting Point: 97-101° Beilstein Reference: 1363961

Formula: C₇H₆O₃
Boiling Pt: 60…65 °C (12 mmHg)
Melting Pt: 97…101 °C
Density: 0,859 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003333
CAS Number: 1194-98-5
EINECS: 214-789-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR