4-Methoxybenzyl alcohol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Anisyl alcohol, p-Anisyl Alcohol, 4-(Hydroxymethyl)anisole, Anise alcohol, (4-Methoxyphenyl)methanol
A15559-0E
A15559-22
A15559-36
CAAAA15559-0EEA
843.72
CAD
CAAAA15559-0E
CAAAA15559-22
CAAAA15559-36
4-Methoxybenzyl alcohol 98%
4-Methoxybenzyl alcohol
Purity: 98%
CAS No.: 105-13-5
Molecular Formula: C8H10O2
Formula Weight: 138.17
Storage Temperature: 2°C to 8°C
Flash Point: 113°(235°F)
Refractive Index: 1.5450
Melting Point: 22-25°
Boiling Point: 259°
Beilstein Reference: 636654
Merck Reference: 14,604
MDL No.: MFCD00004653
Formula:
C₈H₁₀O₂ MW: 138.17 g/mol Boiling Pt: 145 Melting Pt: 22…25 °C Density: 1.110 Flash Pt: 113 °C (235 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00004653 CAS Number: 105-13-5 EINECS: 203-273-6 Merck Index: 14,604 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...