4-Methylveratrole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15558-06
A15558-14
CAAAA15558-06EA
75.21
CAD
CAAAA15558-06
CAAAA15558-14
4-Methylveratrole 98%
4-Methylveratrole
Purity: 98%
CAS No.: 494-99-5
Molecular Formula: C9H12O2
Formula Weight: 152.19
Flash Point: 84°(183°F)
Refractive Index: 1.5280
Odor: Aromatic
Melting Point: 25-28°
Boiling Point: 220°
Beilstein Reference: 2045328
MDL No.: MFCD00016651
Formula:
C₉H₁₂O₂ Boiling Pt: 220 °C Melting Pt: 25…28 °C Density: 1.059 Flash Pt: 84 °C(183 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00016651 CAS Number: 494-99-5 EINECS: 207-796-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...