Ethyl chloroacetate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15554-0E
A15554-22
A15554-36
CAAAA15554-0EEA
176.96
CAD
CAAAA15554-0E
CAAAA15554-22
CAAAA15554-36
Ethyl chloroacetate 99%
Ethyl chloroacetate
Purity: 99%
CAS No.: 105-39-5
Molecular Formula: C4H7ClO2
Formula Weight: 122.55
Flash Point: 54°(149°F)
Refractive Index: 1.4210
Odor: Pungent
Melting Point: -26°
Boiling Point: 142-144°
Beilstein Reference: 506455
Merck Reference: 14,3783
UN No.: 1181
MDL No.: MFCD00000932
Formula:
C₄H₇ClO₂ MW: 122.55 g/mol Boiling Pt: 142…144 °C Melting Pt: –26 °C Density: 1.159 Flash Pt: 54 °C (149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000932 CAS Number: 105-39-5 EINECS: 203-294-0 UN: 1181 ADR: 6.1,II Merck Index: 14,3783 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...