2-Thiophenesulfonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15553-06
A15553-14
CAAAA15553-06EA
72.21
CAD
CAAAA15553-06
CAAAA15553-14
CAAAA15553-22
2-Thiophenesulfonyl chloride 97%
2-Thiophenesulphonyl chloride
Purity: 97%
CAS No.: 16629-19-9
Molecular Formula: C4H3ClO2S2
Formula Weight: 182.65
Flash Point:
Formula:
C₄H₃ClO₂S₂ Boiling Pt: 80…81 °C/1 mm Melting Pt: 29…31 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005426 CAS Number: 16629-19-9 EINECS: 240-677-1 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...