Propylene carbonate 99%

Supplier: Thermo Scientific Chemicals
Warning

A15552-0B A15552-30
CAAAA15552-0BEA 106.97 CAD
CAAAA15552-0B CAAAA15552-30
Propylene carbonate 99%
Propylene carbonate

Purity: 99% CAS No.: 108-32-7 Molecular Formula: C4H6O3 Formula Weight: 102.09 Flash Point: 132°(270°F) Physical Form: Liquid Density: 1.204 Refractive Index: 1.4210 Melting Point: -49° Boiling Point: 240-243° MDL No.: MFCD00005385

Formula: C₄H₆O₃
Boiling Pt: 240…243 °C
Melting Pt: –49 °C
Density: 1.204
Flash Pt: 132 °C(270 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005385
CAS Number: 108-32-7
EINECS: 203-572-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR