5-Bromopentyl acetate 98%

Supplier: Thermo Scientific Chemicals

A15549-06 A15549-14 A15549-22
CAAAA15549-06EA 44.24 CAD
CAAAA15549-06 CAAAA15549-14 CAAAA15549-22
5-Bromopentyl acetate 98%
5-Bromopentyl acetate

Purity: 98% CAS No.: 15848-22-3 Molecular Formula: CH3CO2(CH2)5Br Formula Weight: 209.09 Flash Point: 110°(230°F) Refractive Index: 1.4620 Boiling Point: 109-110°/15mm Beilstein Reference: 1752109 MDL No.: MFCD00039204

Formula: C₇H₁₃BrO₂
Boiling Pt: 109…110 °C/15 mm
Density: 1.255
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039204
CAS Number: 15848-22-3
EINECS: 239-961-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR