Pyrimidine 99%

Supplier: Thermo Scientific Chemicals
Warning

A15534-03 A15534-06 A15534-14
CAAAA15534-03EA 59.88 CAD
CAAAA15534-03 CAAAA15534-06 CAAAA15534-14
Pyrimidine 99%
Pyrimidine

Purity: 99% CAS No.: 289-95-2 Molecular Formula: C4H4N2 Formula Weight: 80.09 Flash Point: 31°(87°F) Refractive Index: 1.5040 Melting Point: 20-22° Boiling Point: 123-124° Beilstein Reference: 103894 Merck Reference: 14,7987 UN No.: 1993 MDL No.: MFCD00006059

Formula: C₄H₄N₂
Boiling Pt: 123…124 °C
Melting Pt: 20…22 °C
Density: 1.016
Flash Pt: 31 °C (87 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006059
CAS Number: 289-95-2
EINECS: 206-026-0
UN: 1993
ADR: 3,III
Merck Index: 14,7987

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR