4'-(Trifluoromethyl)acetophenone 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15529-06
A15529-14
CAAAA15529-06EA
92.27
CAD
CAAAA15529-06
CAAAA15529-14
CAAAA15529-22
4'-(Trifluoromethyl)acetophenone 98+%
4'-(Trifluoromethyl)acetophenone
Purity: 98+%
CAS No.: 709-63-7
Molecular Formula: C9H7F3O
Formula Weight: 188.15
Flash Point: 84°(183°F)
Refractive Index: 1.4620
Odor: Characteristic
Melting Point: 29-35°
Boiling Point: 83-85°/9mm
Beilstein Reference: 1870425
MDL No.: MFCD00000401
Formula:
C₉H₇F₃O Boiling Pt: 83…85 °C/9 mm Melting Pt: 29…35 °C Density: 0.924 Flash Pt: 84 °C (183 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000401 CAS Number: 709-63-7 EINECS: 211-913-0 UN: 1325 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...