2,6-Difluoropyridine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15522-06
A15522-14
A15522-22
CAAAA15522-06EA
63.04
CAD
CAAAA15522-06
CAAAA15522-14
CAAAA15522-22
2,6-Difluoropyridine 98+%
2,6-Difluoropyridine
Purity: 98+%
CAS No.: 1513-65-1
Molecular Formula: C5H3F2N
Formula Weight: 115.08
Flash Point: 33°(91°F)
Refractive Index: 1.4370
Odor: Characteristic
Boiling Point: 123-124°
Beilstein Reference: 1422549
UN No.: 1993
MDL No.: MFCD00006227
Formula:
C₅H₃F₂N Boiling Pt: 123…124 °C Density: 1.268 Flash Pt: 33 °C (91 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006227 CAS Number: 1513-65-1 EINECS: 216-152-8 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...