Methyl-4-bromobenzoate 98+%

Supplier: Thermo Scientific Chemicals
Warning

A15517-06 A15517-14 A15517-22
CAAAA15517-06EA 92.27 CAD
CAAAA15517-06 CAAAA15517-14 CAAAA15517-22
Methyl-4-bromobenzoate 98+%
Methyl 4-bromobenzoate

Purity: 98+% CAS No.: 619-42-1 Molecular Formula: C8H7BrO2 Formula Weight: 215.05 Melting Point: 77-81° Beilstein Reference: 2045132 Merck Reference: 14,1408 MDL No.: MFCD00013531

Formula: C₈H₇BrO₂
Boiling Pt: 246 °C (1013 hPa)
Melting Pt: 77…81 °C
Density: 1.69
Storage Temperature: Ambient
MDL Number: MFCD00013531
CAS Number: 619-42-1
EINECS: 210-596-6
Merck Index: 14,1408

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR