Phthalide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15516-22
A15516-36
CAAAA15516-0EEA
1089.79
CAD
CAAAA15516-0E
CAAAA15516-22
CAAAA15516-36
Phthalide 98+%
Phthalide
Purity: 98+%
CAS No.: 87-41-2
Molecular Formula: C8H6O2
Formula Weight: 134.13
Flash Point: 152°(305°F)
Melting Point: 72-75°
Boiling Point: 289-291°
Beilstein Reference: 114632
MDL No.: MFCD00005906
Formula:
C₈H₆O₂ Boiling Pt: 289…291 °C Melting Pt: 72…75 °C Density: 1.1 Flash Pt: 152 °C(305 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005906 CAS Number: 87-41-2 EINECS: 201-744-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...