4-Bromothioanisole 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromophenyl methyl sulfide, 1-Bromo-4-(methylthio)benzene, 4-Bromophenyl methyl sulphide

A15504-14 A15504-22 A15504-36
CAAAA15504-14EA 131.14 CAD
CAAAA15504-14 CAAAA15504-22 CAAAA15504-36
4-Bromothioanisole 98+%
4-Bromothioanisole

Purity: 98+% CAS No.: 104-95-0 Molecular Formula: C7H7BrS Formula Weight: 203.11 Flash Point: 110°(230°F) Odor: Stench Boiling Point: 250° Beilstein Reference: 1906693 MDL No.: MFCD00000102

Formula: C₇H₇BrS
Boiling Pt: 250 °C
Melting Pt: 37…40 °C
Density: 1.4584 g/cm³ (39 °C)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000102
CAS Number: 104-95-0
EINECS: 203-255-8
UN: 3335
ADR: 9,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR