Phenetole 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15501-0F
A15501-AE
A15501-AP
CAAAA15501-0FEA
420.28
CAD
CAAAA15501-0F
CAAAA15501-AE
CAAAA15501-AP
Phenetole 98+%
Phenetole
Purity: 98+%
CAS No.: 103-73-1
Molecular Formula: C8H10O
Formula Weight: 122.17
Flash Point: 57°(134°F)
Refractive Index: 1.5070
Odor: Aromatic
Melting Point: -30°
Boiling Point: 169-170°
Beilstein Reference: 636270
Merck Reference: 14,7229
UN No.: 3271
MDL No.: MFCD00009090
Formula:
C₆H₅OC₂H₅ Boiling Pt: 169…170 °C Melting Pt: –30 °C Density: 0.967 Flash Pt: 57 °C (134 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009090 CAS Number: 103-73-1 EINECS: 203-139-7 UN: 3271 ADR: 3,III Merck Index: 14,7229 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...