2-Nitrobenzyl alcohol 97%

Supplier: Thermo Scientific Chemicals
Warning

A15496-14 A15496-22 A15496-36
CAAAA15496-14EA 93.85 CAD
CAAAA15496-14 CAAAA15496-22 CAAAA15496-36
2-Nitrobenzyl alcohol 97%
2-Nitrobenzyl alcohol

Purity: 97% CAS No.: 612-25-9 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Melting Point: 70-74° Boiling Point: 270° MDL No.: MFCD00007186

Formula: C₇H₇NO₃
Boiling Pt: 270 °C
Melting Pt: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00007186
CAS Number: 612-25-9
EINECS: 210-302-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR