4-Chloro-1,3-difluorobenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15495-09
A15495-18
A15495-30
CAAAA15495-09EA
42.98
CAD
CAAAA15495-09
CAAAA15495-18
CAAAA15495-30
4-Chloro-1,3-difluorobenzene 98%
4-Chloro-1,3-difluorobenzene
Purity: 98%
CAS No.: 1435-44-5
Molecular Formula: C6H3ClF2
Formula Weight: 148.54
Flash Point: 32°(89°F)
Refractive Index: 1.4750
Odor: Aromatic
Melting Point: -26°
Boiling Point: 126-127°
Beilstein Reference: 2081077
UN No.: 1993
MDL No.: MFCD00042569
Formula:
C₆H₃ClF₂ Boiling Pt: 126…127 °C Melting Pt: –26 °C Density: 1.353 Flash Pt: 32 °C(89 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042569 CAS Number: 1435-44-5 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...