2-Chloro-4-fluoroaniline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15492-06
A15492-14
A15492-22
CAAAA15492-06EA
60.51
CAD
CAAAA15492-06
CAAAA15492-14
CAAAA15492-22
2-Chloro-4-fluoroaniline 97%
2-Chloro-4-fluoroaniline
Purity: 97%
CAS No.: 2106-02-7
Molecular Formula: C6H5ClFN
Formula Weight: 145.56
Flash Point: 98°(208°F)
Refractive Index: 1.5540
Odor: Characteristic
Boiling Point: 191-192°
Beilstein Reference: 2802559
UN No.: 2810
MDL No.: MFCD00042530
Formula:
C₆H₅ClFN Boiling Pt: 191…192 °C Density: 1.219 Flash Pt: 98 °C(208 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042530 CAS Number: 2106-02-7 EINECS: 218-282-0 UN: 2810 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...