Tetrabutylammonium iodide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tetra-n-butylammonium iodide, TBAI

A15484-0B A15484-18 A15484-30
CAAAA15484-0BEA 619.36 CAD
CAAAA15484-0B CAAAA15484-18 CAAAA15484-30
Tetrabutylammonium iodide 98%
Tetrabutylammonium iodide

Purity: 98% CAS No.: 311-28-4 Molecular Formula: [CH3(CH2)3]4NI Formula Weight: 369.38 Melting Point: 142-148° Beilstein Reference: 3916152

Formula: C₁₆H₃₆IN
MW: 369.37 g/mol
Melting Pt: 142…148 °C
Density: 1.0027 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011636
CAS Number: 311-28-4
EINECS: 206-220-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR