4-Fluoro-m-toluidine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15482-06
A15482-14
CAAAA15482-06EA
94.33
CAD
CAAAA15482-06
CAAAA15482-14
4-Fluoro-m-toluidine 97%
4-Fluoro-m-toluidine
Purity: 97%
CAS No.: 452-69-7
Molecular Formula: C7H8FN
Formula Weight: 125.15
Flash Point: 95°(203°F)
Refractive Index: 1.5370
Odor: Characteristic
Melting Point: 32-36°
Boiling Point: 85-86°/9mm
Beilstein Reference: 2935561
UN No.: 2811
MDL No.: MFCD00025294
Formula:
C₇H₈FN Boiling Pt: 85…86 °C/9 mm Melting Pt: 32…36 °C Density: 1.120 Flash Pt: 95 °C (203 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00025294 CAS Number: 452-69-7 EINECS: 207-207-7 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...