3-Nitrobenzamide 98%

Supplier: Thermo Scientific Chemicals
Warning

A15481-09 A15481-18 A15481-30
CAAAA15481-09EA 38.55 CAD
CAAAA15481-09 CAAAA15481-18 CAAAA15481-30
3-Nitrobenzamide 98%
3-Nitrobenzamide

Purity: 98% CAS No.: 645-09-0 Molecular Formula: C7H6N2O3 Formula Weight: 166.14 Melting Point: 140-143° Beilstein Reference: 777185

Formula: C₇H₆N₂O₃
MW: 166.14 g/mol
Boiling Pt: 310
Melting Pt: 140…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00007984
CAS Number: 645-09-0
EINECS: 211-431-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR