4-Methylbenzophenone 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
p-Methylbenzophenone, Phenyl(p-tolyl)methanone
A15479-18
A15479-36
CAAAAA15479-18EA
119.76
CAD
CAAAAA15479-18
CAAAAA15479-30
CAAAA15479-36
4-Methylbenzophenone 98%
4-Methylbenzophenone
Purity: 98%
CAS No.: 134-84-9
Molecular Formula: C14H12O
Formula Weight: 196.25
Flash Point: 143°(289°F)
Melting Point: 53-57°
Boiling Point: 326°
Beilstein Reference: 1909310
Merck Reference: 14,7317
MDL No.: MFCD00008553
Formula:
C₁₄H₁₂O Boiling Pt: 326 °C Melting Pt: 53…57 °C Density: 0.9926 g/cm³ (20 °C) Flash Pt: 143 °C(289 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008553 CAS Number: 134-84-9 EINECS: 205-159-1 Merck Index: 14,7317 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...