2-Chloro-4-fluorobenzonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

A15478-06 A15478-14
CAAAA15478-06EA 110.76 CAD
CAAAA15478-06 CAAAA15478-14
2-Chloro-4-fluorobenzonitrile 98%
2-Chloro-4-fluorobenzonitrile

Purity: 98% CAS No.: 60702-69-4 Molecular Formula: C7H3ClFN Formula Weight: 155.56 Melting Point: 62-66° Beilstein Reference: 6323034 UN No.: 3439 MDL No.: MFCD00042523

Formula: C₇H₃ClFN
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 62…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00042523
CAS Number: 60702-69-4
EINECS: 262-384-8
UN: 3439
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR