4-Nitro-2,6-xylenol 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Hydroxy-5-nitro-3-xylene, 2,6-Dimethyl-4-nitrophenol

A15472-06 A15472-14
CAAAA15472-06EA 90.69 CAD
CAAAA15472-06 CAAAA15472-14
4-Nitro-2,6-xylenol 98%
4-Nitro-2,6-xylenol

Purity: 98% CAS No.: 2423-71-4 Molecular Formula: C8H9NO3 Formula Weight: 167.17 Melting Point: ca 168° dec. Beilstein Reference: 1873275

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Melting Pt: ∼ 168 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007339
CAS Number: 2423-71-4
EINECS: 219-353-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR