3-Methoxybenzyl alcohol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(Hydroxymethyl)anisole, (3-Methoxyphenyl)methanol
A15471-14
A15471-22
CAAAA15471-14EA
88.48
CAD
CAAAA15471-14
CAAAA15471-22
3-Methoxybenzyl alcohol 98+%
3-Methoxybenzyl alcohol
Purity: 98+%
CAS No.: 6971-51-3
Molecular Formula: C8H10O2
Formula Weight: 138.17
Flash Point: 109°(228°F)
Refractive Index: 1.5440
Boiling Point: 252-254°
Beilstein Reference: 2042063
MDL No.: MFCD00004637
Formula:
C₈H₁₀O₂ MW: 138.17 g/mol Boiling Pt: 250 Melting Pt: 29…30 °C Density: 1.112 Flash Pt: 109 °C (228 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004637 CAS Number: 6971-51-3 EINECS: 230-200-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...