Diethyl malonate 99%
Supplier: Thermo Scientific Chemicals
Warning
|
A15468-0C
A15468-0E
A15468-36
CAAAA15468-0CEA
597.24
CAD
CAAAA15468-0C
CAAAA15468-0E
CAAAA15468-36
Diethyl malonate 99%
Diethyl malonate
Purity: 99%
CAS No.: 105-53-3
Molecular Formula: C7H12O4
Formula Weight: 160.17
Flash Point: 90°(194°F)
Physical Form: Liquid
Density: 1.055
Refractive Index: 1.4140
Odor: Pleasant
Melting Point: -50°
Boiling Point: 198-200°
Beilstein Reference: 774687
Merck Reference: 14,3823
MDL No.: MFCD00009195
Formula:
C₇H₁₂O₄ Boiling Pt: 198…200 °C Melting Pt: –50 °C Density: 1.055 Flash Pt: 90 °C(194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009195 CAS Number: 105-53-3 EINECS: 203-305-9 Merck Index: 14,3823 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...