4-(Trifluoromethoxy)benzenesulfonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(Trifluoromethoxy)benzene-1-sulfonyl chloride, 4-(Trifluoromethoxy)benzene-1-sulphonyl chloride

A15467-03 A15467-06 A15467-14
CAAAA15467-03EA 54.83 CAD
CAAAA15467-03 CAAAA15467-06 CAAAA15467-14
4-(Trifluoromethoxy)benzenesulfonyl chloride 98%
4-(Trifluoromethoxy)benzenesulphonyl chloride

Purity: 98% CAS No.: 94108-56-2 Molecular Formula: C7H4ClF3O3S Formula Weight: 260.62 Flash Point: 110°(230°F) Refractive Index: 1.4800 Boiling Point: 120-122°/15mm Beilstein Reference: 7641524 UN No.: 3265 MDL No.: MFCD00042408

Formula: C₇H₄ClF₃O₃S
MW: 260.62 g/mol
Boiling Pt: 120…122 °C/15 mm
Density: 1.536
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042408
CAS Number: 94108-56-2
EINECS: 302-397-9
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR